Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneJET Genomic DNA Purification Kit
GSA_VA

Catalog No. ferk0722
Click to view available options
Quantity:
250 Preps
50 Preps

Thermo Scientific GeneJET Genomic DNA Purification Kit is designed for rapid and efficient purification of high quality genomic DNA from various mammalian cell culture and tissue samples, whole blood, bacteria and yeast.

Thermo Scientific GeneJET Genomic DNA Purification Kit is designed for rapid and efficient purification of high quality genomic DNA from various mammalian cell culture and tissue samples, whole blood, bacteria and yeast.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting and enzymatic reactions.

Highlights
Universal—can be used for both cell and tissue samples from a wide range of sources
Efficient—high yields of high molecular weight genomic DNA
Fast—20 minute procedure following cell lysis
Convenient—spin columns are capped and assembled with collection tubes

Applications
Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Column Type Spin Column
Content And Storage • 5 x 1.2 mL Proteinase K Solution; store at room temperature until opened, –20°C after opening
• 5 x 1 mL RNase A Solution; store at room temperature until opened, –20°C after opening
• 55 mL Digestion Solution; room temperature
• 2 x 60 mL Lysis Solution; room temperature
• 40 mL Wash Buffer I (concentrated); room temperature
• 40 mL Wash Buffer II (concentrated); room temperature
• 150 mL Elution Buffer; room temperature
• 250 GeneJET Purification Columns with Collection Tubes; room temperature
• 250 Collection Tubes; room temperature

Isolation Technology Silica Spin Column
Scale Mini
Sample Type Bacteria, Cells, Rodent Tail, Tissue, Whole Blood, Yeast, Buccal Samples
Elution Volume 200 μL
Final Product Type gDNA
For Use With (Application) PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput Compatibility Not High-throughput Compatible (Manual)
Quantity 250 Preps
Shipping Condition Room Temperature
Test Time 20 min. (After sample homogenization)
Yield ≤30 μg
Starting Material Amount Bacteria: ≤109 cells
Cells: ≤106
Rodent Tail: 0.6 cm (rat), 0.5 cm (mouse)
Tissue: ≤20 mg
Whole Blood: ≤200 μL
Yeast: ≤108 cells
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.