Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneJET Plant RNA Purification Kit

Catalog No. p-4530780
Click to view available options
Quantity:
250 Preps
50 Preps

Requires

Dithiothreitol (DTT, #R0861)

GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.

Thermo Scientific GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis. Purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis. RNA yields vary between different species, tissues, and age of tissue sample. See Table 1 for typical total RNA yields from various sources.

Highlights

  • Fast—procedure completed in 20 min following cell lysis
  • Convinient—protocol does not include clogging-prone lysate filtration step
  • High yield—up to 65 μg of high purity and high integrity RNA from 50 mg plant sample
  • Compatible with wide variety of samples (leaves, roots, sprouts and other parts of various plants)

Applications

Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including:

  • RT-PCR
  • RT-qPCR
  • Northern blotting
  • Nuclease protection assay
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 40 mL Plant RNA Lysis Solution; store at room temperature
• 40 mL Wash Buffer WB 1 (concentrated); room temperature
• 30 mL Wash Buffer 2 (concentrated); room temperature
• 30 mL Water, Nuclease-free; room temperature
• 50 GeneJET RNA Spin Columns with Collection Tubes; For better long-term performance, it is recommended to store the purification columns at 2°C-8°C.
• 50 Collection Tubes (2 mL); room temperature
• 50 Collection Tubes (1.5 mL); room temperature

Isolation Technology Silica Spin Column
Purification Time 20 min. (After sample homogenization)
Elution Volume 50 μL
Sample Type Plants
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Quantity 50 Preps
Scale Mini
Shipping Condition Room Temperature
Starting Material Amount ≤100 mg
Yield Up to 65 μg
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.