Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneJET RNA Cleanup and Concentration Micro Kit

Catalog No. p-4569681
Click to view available options
Quantity:
250 Preps
50 Preps

GeneJET RNA Cleanup and Concentration Micro Kit is designed for rapid and efficient concentration of prepurified RNA samples, as well as for RNA cleanup after DNase I treatment and other enzymatic reactions.

GeneJET RNA Cleanup and Concentration Micro Kit is designed for rapid and efficient concentration of prepurified RNA samples, as well as for RNA cleanup after DNase I treatment and other enzymatic reactions.

The kit combines the convenience of spin column technology with the selective binding properties of a silica membrane, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.

Thermo Scientific GeneJET RNA Cleanup and Concentration Micro Kit is designed for rapid and efficient concentration of prepurified RNA samples, as well as for RNA cleanup after DNase I treatment and other enzymatic reactions.

The kit combines the convenience of spin column technology with the selective binding properties of a silica membrane, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.

Highlights

  • Advanced column design—easy to open, load and handle.
  • Elute in 6 to 10 μL volume.
  • 85 to 100% recovery of a high integrity RNA.
  • Fast and simple—procedure takes less than 5 minutes.

Applications:

  • The purified RNA can be used in common downstream applications
  • RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.
Format Spin Column
Isolation Technology Spin Column
Purification Time 5 min.
Sample Type RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nuclease Protection Assays, Nucleic Acid Labeling, Northern Blotting, cDNA Library Construction
No. of Reactions 50 Preps
Product Line GeneJET
Quantity 50 Preps
Scale Micro
Target Total RNA
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.