Learn More
Thermo Scientific™ phiX174 DNA/BsuRI (HaeIII) Marker
Description
phiX174 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the markers preparation.
phiX174 DNA/BsuRI (HaeIII) Marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively it can be labeled radioactively or non-radioactively with Klenow Fragment or Klenow Fragment, exo- with the fill-in reaction.
- Sizing and approximate quantification of small DNA fragments
- Sharp bands
- Supplied with loading dye for sample DNA
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Gel Type | Agarose |
| Product Type | DNA/BSURI (HAEIII) Marker |
| Ready to Load | No |
| Gel Compatibility | Agarose Gels |
| Green Features | Sustainable packaging |
| Product Line | phiX174 DNA/BsuRI (HaeIII) Marker |
| Quantity | 50 μg |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.