Learn More
Thermo Scientific™ T4 Polynucleotide Kinase (10 U/μL)
GSA_VA
T4 Polynucleotide Kinase (T4 PNK) catalyzes the transfer of the gamma-phosphate from ATP to the 5'-OH group of oligonucleotides, ss and dsDNA and RNA.
$101.40 - $397.50
Specifications
Concentration 10 U/μL
Enzyme T4 Polynucleotide Kinase
Compatible Buffer 10X Reaction Buffer
Product Type T4 Polynucleotide Kinase
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FEREK0031
GSA_VA
View Documents Promotion Details
Thermo Scientific™
EK0031
500 units
Each for $101.40
Only null left
Add to Cart
FEREK0032
GSA_VA
View Documents
Thermo Scientific™
EK0032
2,500 units
Each for $397.50
Only null left
Edge
Add to Cart
Description
Description
T4 Polynucleotide Kinase (T4 PNK) catalyzes the transfer of the gamma-phosphate from ATP to the 5'-OH group of single- and double-stranded DNAs and RNAs, oligonucleotides or nucleoside 3'-monophosphates (forward reaction). The reaction is reversible. In the presence of ADP T4 Polynucleotide Kinase exhibits 5'-phosphatase activity and catalyzes the exchange of phosphate groups between 5'-P-oligo-polynucleotides and ATP (exchange reaction).
The enzyme is also a 3'-phosphatase.
Highlights
  • Active in Thermo Scientific restriction enzyme, RT, and T4 DNA Ligase buffers
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
10 U/μL
T4 Polynucleotide Kinase
10X Reaction Buffer
T4 Polynucleotide Kinase
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.