Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FERMT2 Mouse anti-Canine, Human, Rat, Clone: OTI9E4, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500504

Description
FERMT2 participates in the connection between ECM adhesion sites and the actin cytoskeleton and also in the orchestration of actin assembly and cell shape modulation. It recruits migfilin (FBLP1) protein to cell-ECM focal adhesion sites.Specifications
FERMT2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96AC1 | |
FERMT2 | |
Full-length protein expressed in 293T cell transfected with human FERMT2 expression vector | |
100 μL | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
OTI9E4 | |
Unconjugated | |
FERMT2 | |
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10979, 289992, 490693 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction