Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FERMT2 Mouse anti-Human, Clone: OTI25A4, liquid, TrueMAB™
SDP

Catalog No. 501671640
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1640 Supplier OriGene Supplier No. TA500507
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FERMT2 participates in the connection between ECM adhesion sites and the actin cytoskeleton and also in the orchestration of actin assembly and cell shape modulation. It recruits migfilin (FBLP1) protein to cell-ECM focal adhesion sites.

Specifications

Antigen FERMT2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone OTI25A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FERMT2
Gene Accession No. Q96AC1
Gene Alias KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2
Gene Symbols FERMT2
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human FERMT2 expression vector
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10979
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.