Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FERMT3/URP2 Antibody (OTI3C7), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272241R
Description
FERMT3/URP2 Monoclonal specifically detects FERMT3/URP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FERMT3/URP2 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FERMT3 | |
Recombinant protein fragment corresponding to amino acids 259-528 of human FERMT3/URP2 (NP_113659) produced in E.coli. | |
0.1 mL | |
83706.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3C7 | |
Western Blot, Immunohistochemistry | |
fermitin family member 3, KIND3fermitin family homolog 3 (Drosophila), kindlin 3, Kindlin-3, MGC10966, MIG-2, MIG2BURP2SF, MIG2-like protein, UNC-112 related protein 2, UNC112C, Unc-112-related protein 2, URP2fermitin family homolog 3 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction