Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferno™ EZ Glide™ Evacuation Chair: Accessories

Click to view available options
Product Type:
Storage Cover
Material:
Vinyl
Description
For Evacuation Chair, see cat. no. 19161597.
Specifications
Specifications
| Product Type | Storage Cover |
| Material | Vinyl |
| For Use With (Equipment) | EZ glide |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction