Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferric Alum Indicator, Volhard, Reagents
Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | Ferric Alum Indicator, Volhard |
CAS | 7732-18-5, 7697-37-2 |
Boiling Point | 102°C |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.08 |
Color | Amber |
pH | <1 |
Quantity | 1 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction