Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ferric Antioxidant Status Detection Kit

Catalog No. EIAFECL2
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAFECL2 2 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIAFECL2 Supplier Invitrogen™ Supplier No. EIAFECL2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

96-well plates, standard, other assay specific reagents, buffers and lot-specific technical data sheet.

Assay Ferric Antioxidant Power (FRAP) in Serum, Plasma, Urine, Teas, Fruit Juices, Beer, Cider, Cell Lysates, Herbal and Fruit Extracts.

The Ferric Antioxidant Status Detection research-use-only kit is a colorimetric assay designed for the quantification and detection of Ferric Antioxidant Status (also referred as Ferric Reducing Antioxidant Power, FRAP) in serum, plasma, urine, teas, fruit juices, beer, cider, cell lysates, herbal and fruit extracts.

This complete, ready-to-use kit includes clear 96-well plate(s), ferrous chloride standard, ascorbic acid control, and other components to perform the assay. A 96-well microplate reader capable of reading optical density at 560 nm is required for use of this kit.

Performance characteristics
• Assay type: colorimetric detection kit
• Sample types: serum, plasma, urine, teas, fruit juices, beer, cider, cell lysates, herbal and fruit extracts
• Sensitivity: 8.06 μM
• Standard curve range: 31.2–1000 μM
• Reactivity: species independent

Background
Reactive oxygen species (ROS) are produced as a consequence of normal aerobic metabolism. These free radicals (FR) are usually removed or converted into other products in vivo by an array of antioxidants. Organisms have developed complex antioxidant systems to protect themselves from oxidative stress, including the activity of specific enzymes (especially superoxide dismutase, glutathione peroxidase, catalase, glutathione reductase) as well as nonenzymatic compounds with antioxidant activity, such as beta-tocopherol, L-ascorbic acid, glutathione, coenzyme Q10, flavonoids, and albumin. The assay measures the antioxidant ability from all species.

Assay principle
The Ferric Antioxidant Status Detection kit is designed to quantitatively measure antioxidant status in a variety of samples. A ferrous chloride standard is provided to generate a standard curve for the assay and all samples should be read off of the standard curve. Samples are diluted in assay buffer and added to the wells. The FRAP color solution is made by mixing reagent A and B with assay buffer. The FRAP color solution is added to all wells and the plate incubated at room temperature. Antioxidant power in the samples reacts with the FRAP color solution to generate a blue-colored product which is read at 560 nm.


TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 96-well plate(s), standard, other assay specific reagents, buffers, and lot-specific technical data sheet.

Store at –20°C.
Detection Method Colorimetric
Immunoassay Kit Format Colorimetric Detection Kit
Product Type Ferric Antioxidant Status Detection Kit
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Equipment) Microplate Reader
Gene ID (Entrez) Non-Gene
Gene Symbol Non-Gene
Quantity 2 x 96 Tests
Shipping Condition Approved for shipment on Wet or Dry Ice
Target Kit Named Ferric Antioxidant Status
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.