Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Benzoylacetonate ca 11% Fe, Thermo Scientific™

Catalog No. AC410540100
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14323-17-2
Molecular Formula C30H27FeO6
MDL Number MFCD00041938
Synonym Tris(1-phenyl-1, 3-butanediono)iron(III)

Specifications

Color Red to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10 g
Formula Weight 539.39
Percent Purity ∼11% Fe, certified metal content supplied with order
Physical Form Powder
Chemical Name or Material Ferric benzoylacetonate, ca. 11% Fe
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.