Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride, 100% (w/v) (100 g + 40 mL Water), Ricca Chemical
SDP

Catalog No. 313016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3130-16 500 mL Natural Poly Bottle
3130-4 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3130-16 Supplier Ricca Chemical Company Supplier No. 313016
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Amber
Percent Purity 100%
Grade Laboratory
Packaging Natural Poly Bottle
pH Acidic
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 100% (w/v) Aqueous Solution (100g + 40mL Water)
Chemical Name or Material Ferric Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.