Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferric Chloride Acid Solution, Reagents
Click to view available options
:
Quantity: 100 mL
Specifications
Specifications
| Chemical Name or Material | Ferric Chloride Acid Solution |
| Color | Orange/Brown |
| CAS | 7732-18-5, 7697-37-2 |
| pH | <1 |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Synonym | Iron(III) Chloride |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.0728 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction