Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride Oxidizing Solution, 1% (w/v), with 1.6% (w/v) Sulfamic Acid, Ricca Chemical
SDP

Catalog No. p-3251669
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle

For Cyanide Determination

Chemical Identifiers

CAS 10025-77-1, 5329-14-6, 7732-18-5
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Color Green-Yellow
Percent Purity 1%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous,with 1.6% (w/v) Sulfamic Acid
pH Acidic
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 1% (w/v)
Chemical Name or Material Ferric Chloride Oxidizing Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.