Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ferric Nitrate, Nonahydrate
SCI_ED

Catalog No. s25319 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
IUPAC Name iron(3+) nonahydrate trinitrate
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Gray to White or Purple
Physical Form Solid
Quantity 100 g
Assay Percent Range >98%
Linear Formula Fe(NO3)3 · 9H2O
Formula Weight 404
Grade Reagent
Packaging Reagent Grade
Chemical Name or Material Ferric Nitrate, Nonahydrate
ShelfLife 1 year
DOTInformation DOT Class 5.1, III, : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.