Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ferric Nitrate Solution, 1M
SCI_ED

Catalog No. s25320 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
Packaging:
Reagent Grade

Catalog No. S25320

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
IUPAC Name iron(3+) nonahydrate trinitrate
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Quantity 500 mL
Assay Percent Range >98%
Linear Formula Fe(NO3)3·9H2O
Formula Weight 404
Grade Reagent
Packaging Reagent Grade
Density 1.68g/mL
Chemical Name or Material Ferric Nitrate Solution, 1M
ShelfLife 1 year
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.