Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ferric(ous) Oxide, Black
SCI_ED

Catalog No. s25321 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Lab Grade
Used to test magnetic properties

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 1317-61-9
Molecular Formula Fe3O4
Molecular Weight (g/mol) 231.53
MDL Number MFCD00011010
InChI Key WTFXARWRTYJXII-UHFFFAOYSA-N
Synonym Black Iron Oxide
IUPAC Name diiron(3+) λ²-iron(2+) tetraoxidandiide
SMILES [O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]

Specifications

Quantity 500 g
Linear Formula Fe3O4
Formula Weight 231.54
Grade Laboratory
Packaging Lab Grade
Density 5.18g/mL
Chemical Name or Material Ferric(ous) Oxide, Black
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.