Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferric Oxide, Red, Powder, Technical, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Ferric Oxide, Red, Powder, Technical , also known as iron(II) oxide, is one of the main oxides of iron. It is sometimes referred to as rust and also occurs naturally as the mineral magnetite. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 2.5 kg |
| Grade | Technical |
| Packaging | Amber Glass Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction