Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Subsulfate Solution, USP, 20-22 g/100 mL, Spectrum™ Chemical
SDP

Catalog No. 18602680 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
6 x 500 mL
200 L
Packaging:
Amber Glass Bottle
Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-680 500 mL Amber Glass Bottle
18-602-681 6 x 500 mL Amber Glass Bottle
18-602-682 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-680 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F1305500ML
Add to Cart
Add to Cart

F1305, 1310-45-8, Fe4(OH)2(SO4)5

Spectrum™ Chemical Ferric Subsulfate Solution, USP All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1310-45-8
Molecular Formula Fe4H2O22S5
Molecular Weight (g/mol) 737.67
InChI Key GDPKWKCLDUOTMP-UHFFFAOYSA-B
IUPAC Name tetrairon(3+) dihydroxide pentasulfate
SMILES [OH-].[OH-].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 mL
Grade USP
Concentration or Composition (by Analyte or Components) 20-22 g/100 mL
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.