Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ferric Sulfate, Hydrated
SCI_ED

Catalog No. s25322 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10028-22-5
Molecular Formula Fe2O12S3
Molecular Weight (g/mol) 399.858
InChI Key RUTXIHLAWFEWGM-UHFFFAOYSA-H
Synonym ferric sulfate, iron iii sulfate, iron persulfate, iron tersulfate, diiron tris sulphate, diiron trisulfate, ferric persulfate, ferric tersulfate, iron sesquisulfate, ferric sesquisulfate
PubChem CID 24826
ChEBI CHEBI:53438
IUPAC Name iron(3+);trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3]

Specifications

Color Gray-Yellow
Quantity 100 g
Linear Formula FeSO4 · H2O
Formula Weight 399.87
Grade Reagent
Packaging Reagent Grade
Density 3.097g/mL
Chemical Name or Material Ferric Sulfate, Hydrated
ShelfLife 1 year
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.