Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferritin Antibody (F23), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1108384UV
Description
Ferritin Monoclonal specifically detects Ferritin in Human samples. It is validated for ELISA, Sandwich ELISA.Specifications
Ferritin | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FTH1 | |
This clone has been derived from hybridization of SP2/0 myeloma cells with spleen cells of Balb/c mice immunised with purified human ferritin. | |
0.2 mL | |
Cancer, Cellular Markers, Lipid and Metabolism | |
2495 | |
Human | |
Purified |
ELISA | |
F23 | |
ELISA, Sandwich ELISA | |
apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Ferritin (F23) | |
Store at 4C in the dark. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction