Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferritin Antibody (FTH/2081) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308581
Description
Ferritin Monoclonal specifically detects Ferritin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
| Ferritin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1-2 ug/mL, Immunohistochemistry-Paraffin : 0.1-0.2 ug/mL | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| Recombinant human Ferritin protein fragment (around aa 58-180) (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Cellular Markers, Lipid and Metabolism | |
| 2495 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. | |
| IgG2c κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| FTH/2081 | |
| Unconjugated | |
| 10mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction