Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferritin Antibody (FTH/2082), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308613V
Description
Ferritin Monoclonal antibody specifically detects Ferritin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Ferritin | |
Monoclonal | |
DyLight 405 | |
apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
Recombinant human Ferritin protein fragment (around aa 58-180) (exact sequence is proprietary) (Uniprot: P02794) | |
100 μg | |
Cancer, Cellular Markers, Lipid and Metabolism | |
2495 | |
Store at 4°C in the dark. | |
IgG2c κ |
Western Blot, Immunohistochemistry (Paraffin) | |
FTH/2082 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction