Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferritin Light Chain Antibody (FTL/1386), Novus Biologicals™

Mouse Monoclonal Antibody
$256.00
Specifications
Antigen | Ferritin Light Chain |
---|---|
Clone | FTL/1386 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.1 - 0.2 ug/ml, Flow Cytometry 0.1 - 0.2 ug/million cells, Immunocytochemistry/Immunofluorescence 0.1 - 0.2 ug/ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25329520
![]() |
Novus Biologicals
NBP25329520UG |
20 μg |
Each for $256.00
|
|
|||||
NB253295200
![]() |
Novus Biologicals
NBP253295200UG |
200 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Ferritin Light Chain Monoclonal specifically detects Ferritin Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Ferritin Light Chain | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
2512 | |
Recombinant human FTL protein fragment (aa 38-165) (exact sequence is proprietary) | |
Primary | |
Store at 4C. |
FTL/1386 | |
Western Blot 0.1 - 0.2 ug/ml, Flow Cytometry 0.1 - 0.2 ug/million cells, Immunocytochemistry/Immunofluorescence 0.1 - 0.2 ug/ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Ferritin L subunit, ferritin L-chain, ferritin light chain, ferritin light polypeptide-like 3, ferritin, light polypeptide, MGC71996, NBIA3 | |
FTL | |
IgG2b | |
Protein A or G purified | |
22 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title