Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferritin Light Chain Antibody (FTL/3872R), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308623G
Description
Ferritin Light Chain Monoclonal antibody specifically detects Ferritin Light Chain in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Ferritin Light Chain | |
Monoclonal | |
DyLight 488 | |
Ferritin L subunit, ferritin L-chain, ferritin light chain, ferritin light polypeptide-like 3, ferritin, light polypeptide, MGC71996, NBIA3 | |
Recombinant full-length human Ferritin Light Chain protein (Uniprot: P02792) | |
100 μg | |
Cancer | |
2512 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
FTL/3872R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction