Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ferritin Light Chain Polyclonal Antibody

Rabbit Polyclonal Antibody
$249.00 - $536.00
Specifications
| Antigen | Ferritin Light Chain |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119112
![]() |
Invitrogen™
PA5119112 |
200 μL |
Each for $536.00
|
|
|||||
|
PIPA5121998
|
Invitrogen™
PA5121998 |
20 μL |
Each for $249.00
|
|
|||||
Description
Ferritin Light Chain Polyclonal Antibody for IHC (P), Western Blot
This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes.Specifications
| Ferritin Light Chain | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Ftl1 | |
| ferritin L subunit; Ferritin L subunit 1; ferritin L-chain; ferritin light chain; Ferritin light chain 1; ferritin light polypeptide-like 3; ferritin, light polypeptide; fril; Ftl; Ftl1; Ftl-1; L-ferritin; LFTD; NBIA3 | |
| Ftl1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P29391 | |
| 14325 | |
| Recombinant protein Ferritin, Light Polypeptide. The antigen corresponds to amino acid range 5-183 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title