Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferritin mitochondrial Antibody (SR1345), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321775100UL
Description
Ferritin mitochondrial Monoclonal antibody specifically detects Ferritin mitochondrial in Human samples. It is validated for Western BlotSpecifications
Ferritin mitochondrial | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1345 | |
Western Blot 1:500-1:2000 | |
EC 1.16.3.1, ferritin H subunit, ferritin mitochondrial, ferritin, mitochondrial, mitochondrial ferritin, MTF | |
A synthesized peptide derived from human Ferritin mitochondrial (Uniprot #: Q8N4E7) | |
100 μg | |
Lipid and Metabolism | |
94033 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction