Learn More
Invitrogen™ Ferritin Monoclonal Antibody (513C10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA514736
Description
MA5-14736 detects Ferritin in ELISA, IP, IF, and RIA applications and shows reactivity to human samples. The MA5-14736 Immunogen is human liver ferritin. MA5-14736 detects ferritin which has a predicted molecular weight of approximately 19kDa. Product MA514736 is a smaller package size of MIF2504 (formerly sold as a Seradyn product).
Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. Ferritin is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in the light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. The ferritin gene has multiple pseudogenes, and ferritin is a ubiquitous and highly conserved protein which plays a major role in iron homeostasis. Ferritin is a holoenzyme shell with a molecular weight of about 450 kDa, and consists of 24 subunits of two types, H(heavy) and L(light), and capable of storing up to 4,500 atoms of ferric iron.
Specifications
Ferritin | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P02792, P02794 | |
FTH1, FTL | |
Human liver ferritin. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Immunocytochemistry | |
513C10 | |
Unconjugated | |
FTL | |
apoferritin; cell proliferation-inducing gene 15 protein; Ferritin H subunit; ferritin heavy chain; ferritin heavy chain 1; Ferritin heavy chain, N-terminally processed; Ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 3; Ferritin subunit H; ferritin, heavy polypeptide 1; ferritin, light polypeptide; FHC; frih; fril; FTH; Fth1; FTHL6; FTL; HFE5; H-ferritin; HFt; L apoferritin; LFTD; MFH; MGC71996; NBIA3; OK/SW-cl.84; PIG15; placenta immunoregulatory factor; PLIF; proliferation-inducing protein 15 | |
Mouse | |
Protein A | |
RUO | |
2495, 2512 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.