Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ferritin Monoclonal Antibody (521)
GREENER_CHOICE

Catalog No. PIMA514693
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514693 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514693 Supplier Invitrogen™ Supplier No. MA514693
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-14693 detects Ferritin in ELISA, IP, IF, and RIA applications and shows reactivity to human samples. The MA5-14693 Immunogen is human liver ferritin. MA5-14693 detects ferritin which has a predicted molecular weight of approximately 19kDa. Product MA514693 is a smaller package size of MIF2502 (formerly sold as a Seradyn product).

Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. Ferritin is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in the light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. The ferritin gene has multiple pseudogenes, and ferritin is a ubiquitous and highly conserved protein which plays a major role in iron homeostasis. Ferritin is a holoenzyme shell with a molecular weight of about 450 kDa, and consists of 24 subunits of two types, H(heavy) and L(light), and capable of storing up to 4,500 atoms of ferric iron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ferritin
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Immunocytochemistry
Classification Monoclonal
Clone 521
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene FTL
Gene Accession No. P02792, P02794
Gene Alias apoferritin; cell proliferation-inducing gene 15 protein; Ferritin H subunit; ferritin heavy chain; ferritin heavy chain 1; Ferritin heavy chain, N-terminally processed; Ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 3; Ferritin subunit H; ferritin, heavy polypeptide 1; ferritin, light polypeptide; FHC; frih; fril; FTH; Fth1; FTHL6; FTL; HFE5; H-ferritin; HFt; L apoferritin; LFTD; MFH; MGC71996; NBIA3; OK/SW-cl.84; PIG15; placenta immunoregulatory factor; PLIF; proliferation-inducing protein 15
Gene Symbols FTH1, FTL
Host Species Mouse
Immunogen Human Liver Ferritin.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2495, 2512
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.