Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferrochelatase Rabbit anti-Human, Mouse, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABS212425UL
Description
Anti-Ferrochelatase, Cat. No. ABS2124, is a rabbit polyclonal antibody that detects Ferrochelatase, mitochondrial. It has been tested for use in Immunoprecipitation and Western Blotting.
Specifications
Ferrochelatase | |
Polyclonal | |
Unconjugated | |
Heme synthase;protoheme ferro-lyase | |
Rabbit | |
∽43 kDa observed; 47.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
25 μL | |
Signaling | |
This polyclonal antibody detects ferrochelatase in human and murine species. It targets the mature form of ferrochelatase. | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Serum |
Immunoprecipitation, Western Blot | |
Please refer to lot specific datasheet. | |
Rabbit polyclonal antiserum with 0.05% sodium azide. | |
FECH | |
Flag-tagged recombinant fragment corresponding to the mature mitochondrial form containing 369 amino acids (aa 55-423) from human Ferrochelatase. | |
Unpurified | |
RUO | |
Primary | |
Human, Mouse | |
Antibodies | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction