Learn More
Description
Used for: detection of intracellular labile Fe(II).
Application: high-throughput assays for high content imaging of subcellular labile Fe(II) in live cells; fluorescent microscopy; studies on iron homeostasis and ferroptosis.
Properties and Photophysical Data: FerroOrange has excellent cell-membrane permeability and low cell toxicity. FerroOrange has no chelating ability. For the fluorescent form, excitation and emission maxima (λ) measured in 50 mM HEPES buffer (pH 7.4, 0.2% DMSO) are 545 nm and 575 nm, respectively; extinction coefficient ε = 73,500 M-1cm-1; quantum yield φ = 0.19.
Please refer to this protocol for further information on how to use this product.
Specifications
Specifications
| Chemical Name or Material | 4-(tert-Butoxycarbonyl)-1-(3'-(4-(tert-butoxycarbonyl)piperazin-1-yl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthen]-6'-yl)piperazine 1-oxide |
| CAS | 2711762-70-6 |
| Quantity | 24 μg |
| Product Line | Tocris |
| Target | Ion Indicators |
| Molecular Formula | C38H44N4O8 |
| Purity | 90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
