Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate, 0.00282 N (0.00282 M), 1 mL = 0.1 mg Cl2, Ricca Chemical
SDP

Catalog No. p-3251682
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
IUPAC Name λ2-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Green
Equivalence 1 mL = 0.1 mg Cl2
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 100 mL
Physical Form Liquid
Concentration or Composition Notes 0.00282N,0.00282M
Chemical Name or Material Ferrous Ammonium Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.