Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ferrous Ammonium Sulfate, Hexahydrate
SCI_ED

Catalog No. S25323 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade - Crystals
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25323 100 g Reagent Grade - Crystals
S25323A 500 g Reagent Grade - Crystals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25323

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Ammonium Iron (II) Sulfate
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Green
Quantity 100 g
Linear Formula Fe(NH4)2(SO4)2·6H2O
Formula Weight 392.13
Packaging Reagent Grade - Crystals
Density 1.865g/mL
Chemical Name or Material Ferrous Ammonium Sulfate, Hexahydrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.