Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Fumarate, USP, 97-101%, Spectrum™ Chemical
SDP

Catalog No. 18602696
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

FE108, 141-01-5, C4H2FeO4

Spectrum™ Chemical Ferrous Fumarate, USP is used primarily to treat iron deficiency anemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 141-01-5
Molecular Formula C4H2FeO4
Molecular Weight (g/mol) 169.90
InChI Key PMVSDNDAUGGCCE-ODZAUARKSA-L
IUPAC Name λ²-iron(2+) (2Z)-but-2-enedioate
SMILES [Fe++].[O-]C(=O)\C=C/C([O-])=O

Specifications

CAS Max % 100%
Quantity 2.5 kg
Percent Purity 97 to 101%
Grade USP
Loss on Drying 1.5%
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.