Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Sulfate Solution (5% Fe), Reagents

Catalog No. 01335287
Click to view available options
:
Quantity: 200 L

Ferrous Sulfate Solution (5% Fe), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ferrous Sulfate Solution (5% Fe)
Boiling Point 102°C
Color Light Green
CAS 7732-18-5, 7664-93-9
pH <7
Physical Form Liquid
Quantity 200 L
Percent Purity 5%
Grade Reagent
Packaging Poly Drum
Specific Gravity 1.1457
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.