Learn More
Trajan Scientific and Medical™ Ferrule SilTite™ Metal Kit for Split / Splitless Injectors
SDP
Catalog No. 073271 Shop All SGE Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Agilent Split/Splitless Injector
Catalog No. For Use With (Equipment)
07-327-1 Agilent Split/Splitless Injector
1 options
Catalog No. 07-327-1 Supplier Trajan Scientific and Medical™ Supplier No. 073271
Delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Specifically designed for connecting capillary columns
  • Connects guard columns to capillary columns
  • Inert and precise glass lined bore
  • Low dead volume
  • Suitable for Agilent GCs
Specifications
For Use With (Equipment) Agilent Split/Splitless Injector
Type Ferrule SilTite Metal Kit
Quantity 1 Each
Diameter (Metric) Inside 0.32 mm