Learn More
Promega FES Kinase Enzyme System, 10microg
Catalog No. PRV1981 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV1981 10 μg
1 options
Catalog No. PRV1981 Supplier Promega Supplier No. V1981
Recombinant full-length human FES was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FES is a proto-oncogene that encodes a protein-tyrosine kinase distinct from c-Src, c-Abl and other nonreceptor tyrosine kinases.
  • Easily Screen and Profile FES Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C