Learn More
Invitrogen™ FES Monoclonal Antibody (5A11G5)

Description
MA5-15353 targets FES in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15353 immunogen is purified recombinant fragment of FES expressed in E. Coli.
Specifications
Specifications
| Antigen | FES |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 5A11G5 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | FES |
| Gene Accession No. | P07332 |
| Gene Alias | AI586313; BB137047; c-fes; EC 2.7.10.2; feline sarcoma (Snyder-Theilen) viral (v-fes)/Fujinami avian sarcoma (PRCII) viral (v-fps) oncogene homolog; feline sarcoma oncogene; feline sarcoma/Fujinami avian sarcoma oncogene homolog; Fes; FES proto-oncogene, tyrosine kinase; FPS; Oncogene FES, feline sarcoma virus; p93c-fes; Proto-oncogene c-Fes; proto-oncogene c-Fps; proto-oncogene tyrosine-protein kinase Fes/Fps; RGD1564385; tyrosine kinase Fps/Fes; tyrosine-protein kinase Fes/Fps; Variant 4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.