Learn More
Invitrogen™ FES Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596171
Description
Immunogen sequence: WQQLQQELTK THSQDIEKLK SQYRALARDS AQAKRKYQEA SKDKDRDKAK DKYVRSLWKL FAHHNRYVLG VRAAQLHHQH HHQLLLPGLL RSLQDLHEEM ACILKEILQE YLEISSLVQD EVVAIHREMA AAAARIQPEA EYQGFLRQYG SAPDVPPCVT FDESLLEEGE PLEPGELQLN ELTVESVQHT LTSVTDELAV ATEMVFRRQE MVTQLQQELR NEEENTHPRE RVQLLGKRQV LQEALQGLQV ALCSQAKLQA QQELLQTKLE HLGPGEPPPV LLLQDDRHST SSSEQEREGG R; Positive Samples: Mouse heart, Mouse brain; Cellular Location: Cell junction, Cell membrane, Cytoplasm, Cytoplasmic side, Cytoplasmic vesicle, Golgi apparatus, Peripheral membrane protein, cytoskeleton, cytosol, focal adhesion.
FES (FPS) is a tyrosine protein kinase that is required for maintenance of cellular transformation. Its chromosomal location is linked to a specific translocation event identified in patients with acute promyelocytic leukemia, and it is also involved in normal hematopoiesis. Alternative splicing results in multiple variants encoding different isoforms.
Specifications
| FES | |
| Polyclonal | |
| Unconjugated | |
| FES | |
| AI586313; BB137047; c-fes; EC 2.7.10.2; feline sarcoma (Snyder-Theilen) viral (v-fes)/Fujinami avian sarcoma (PRCII) viral (v-fps) oncogene homolog; feline sarcoma oncogene; feline sarcoma/Fujinami avian sarcoma oncogene homolog; Fes; FES proto-oncogene, tyrosine kinase; FPS; Oncogene FES, feline sarcoma virus; p93c-fes; Proto-oncogene c-Fes; proto-oncogene c-Fps; proto-oncogene tyrosine-protein kinase Fes/Fps; RGD1564385; tyrosine kinase Fps/Fes; tyrosine-protein kinase Fes/Fps; Variant 4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14159 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16879 | |
| FES | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-420 of human FES (NP_0011372561). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.