Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FeSODB1 Chicken anti-Protozoa, Polyclonal, FabGennix

Catalog No. 502207003
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7003 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7003 Supplier Fabgennix Inc Supplier No. FESODB1101AP
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

FeSODB1 Polyclonal antibody specifically detects FeSODB1 in Protozoa samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen FeSODB1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Alias CGC20_34255; CGC21_20505; FeSODB1; LdCL_320024600 LDHU3_32.2410; Superoxide dismutase
Host Species Chicken
Immunogen Purified peptide from FeSODB1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.