Learn More
Gibco™ Fetal Bovine Serum, qualified, United States

Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
Gibco Sera: Trusted Quality for Consistent Performance
Gibco Sera Category: Value Plus (Performance)
- Use for general cell culture with common cell lines
- Low endotoxin and hemoglobin levels
- Endotoxin level: <10 EU/mL
- Hemoglobin level: <25 mg/dL
- Origin: United States
Gibco Serum Delivers
- ISO 13485 certified, processed in FDA registered facilities
- Triple filtered at 0.1 μm
- Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Order Info
Shipping Condition: Dry Ice

Specifications
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
Description | Fetal Bovine Serum, qualified, One Shot™ format, United States |
Species | Bovine |
Age | Fetal |
Country of Origin | United States |
Form | Liquid |
Format | One Shot |
Product Type | Fetal Bovine Serum |
Quantity | 50 mL |
Serum Treatment | Standard (Sterile-filtered) |
Show More |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.