Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Fetal Bovine Serum, Regular, USDA Approved Origin

Click to view available options
Quantity:
50 mL
Serum Treatment:
Heat-inactivated
None
Description
- Exported from the country of origin accompanied with a Veterinarian Health Certificate and imported under a USDA-APHIS issued import permit.
- True pooled and triple 0.1 micron sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements.

Specifications
Specifications
Content And Storage | -40°C to -10°C |
Species | Bovine |
Country of Origin | United States |
Form | Liquid |
Quantity | 50 mL |
Serum Treatment | Heat-inactivated |
Sterility | Sterile |
Source | USDA Origin |
Heat Inactivated | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction