Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Fetal Bovine Serum, Tet system approved, USDA-approved regions
GSA_VA

Catalog No. p-9247004
Encompass_Preferred
Click to view available options
Quantity:
50 mL
500 mL
Format:
Bottle
One Shot

Tet system-approved FBS is formulated for cell culture applications using tetracycline- and tetracycline derivative-regulated gene expression systems.

Tet system-approved FBS is formulated for cell culture applications using tetracycline- and tetracycline derivative-regulated gene expression systems. Cell-based assays that detect the presence of tetracycline and tetracycline analogs are used to select FBS that delivers the highest range of induction.

  • Designed to ensure the absence of detectable tetracycline and its derivatives that may interfere with accurate regulation of gene expression
  • Functional testing provides confidence that unintended tetracyline gene expression will not result
  • Sera category: specialty

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 13485 Certified
Classification Value
Content And Storage Store at <-10°C.
Species Cattle/Bovine
Cell Type Use for a broad range of cell types, especially sensitive cell lines
Age Fetal
Country of Origin Mexico
Format One Shot
Specialty Type Tet-System Approved
Product Type Fetal Bovine Serum
Quantity 50 mL
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Grade Value
Tested For 50 quality specification tests including 9 CFR virus testing, endotoxin, and performance
Endotoxin Concentration <50 EU/mL (levels routinely <10 EU/mL)
Green Features Sustainable packaging
Hemoglobin Concentration <25 mg/dL
Purity or Quality Grade Value
Product Line One Shot™
Shipping Condition Frozen
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.