Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fetal Hemoglobin Antibody, Alexa Fluor™ 532 , Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals NB11041084AF532
Description
Fetal Hemoglobin Polyclonal antibody specifically detects Fetal Hemoglobin in Human samples. It is validated for Western Blot, ELISASpecifications
Fetal Hemoglobin | |
Polyclonal | |
50mM Sodium Borate | |
Sheep | |
Immunogen affinity purified | |
RUO | |
3047 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 532 | |
abnormal hemoglobin, A-gamma globin, gamma A hemoglobin, gamma globin, gamma-1-globin, gamma-2-globin, gamma-globin chain, G-gamma globin Paulinia, Hb F Agamma, hb F Ggamma, HBGA, HBGR, HBG-T1, HBG-T2, Hemoglobin gamma-1 chain, hemoglobin gamma-2 chain, Hemoglobin gamma-A chain, hemoglobin gamma-G chain, hemoglobin subunit gamma-1, hemoglobin subunit gamma-2, hemoglobin, gamma A, hemoglobin, gamma, regulator of, HSGGL1, methemoglobin, PRO2979 | |
Purified human hemoglobin F of fetal cord blood cell origin. | |
1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction