Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fetal Hemoglobin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11041084 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11041084 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11041084 Supplier Novus Biologicals Supplier No. NB11041084
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody has been used in 5 publications

Fetal Hemoglobin Polyclonal specifically detects Fetal Hemoglobin in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Fetal Hemoglobin
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:30000, ELISA 1:1000 - 1:30000
Formulation PBS with 0.09% Sodium Azide
Gene Accession No. P69892
Gene Alias A-gamma globin, gamma A hemoglobin, gamma globin, gamma-1-globin, Hb F Agamma, HBGA, HBGR, Hemoglobin gamma-1 chain, Hemoglobin gamma-A chain, hemoglobin subunit gamma-1, hemoglobin, gamma A, hemoglobin, gamma, regulator of, HSGGL1
Gene Symbols HBG1
Host Species Sheep
Immunogen Purified human hemoglobin F of fetal cord blood cell origin.
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3047
Test Specificity By immunoelectrophoresis and ELISA this antibody reacts specifically with human hemoglobin F. No antibody was detected against hemoglobin A1, A2, S or human serum proteins.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.