Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fetuin A/AHSG Antibody (1), Alexa Fluor™ 532

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289412AF532
Description
Fetuin A/AHSG Monoclonal specifically detects Fetuin A/AHSG in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA Detection.Specifications
| Fetuin A/AHSG | |
| Monoclonal | |
| Alexa Fluor 532 | |
| A2HS, AHS, alpha-2-HS-glycoprotein, alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, FETUAba-alpha-2-glycoprotein, fetuin-A, HSGA | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Fetuin A/AHSG (NP_001613.2; Met 1-Val 367) . | |
| 0.1 mL | |
| Cancer | |
| 197 | |
| Human | |
| Purified |
| ELISA, Immunofluorescence | |
| 1 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human Cystatin-C Human Cystatin-E Mouse Fetain-A | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction