Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fetuin A/AHSG Antibody (1), DyLight 350

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289412UV
Description
Fetuin A/AHSG Monoclonal specifically detects Fetuin A/AHSG in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA Detection.Specifications
Fetuin A/AHSG | |
Monoclonal | |
DyLight 350 | |
A2HS, AHS, alpha-2-HS-glycoprotein, alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, FETUAba-alpha-2-glycoprotein, fetuin-A, HSGA | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Fetuin A/AHSG (NP_001613.2; Met 1-Val 367) . | |
0.1 mL | |
Cancer | |
197 | |
Human | |
Purified |
ELISA, Immunofluorescence | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human Cystatin-C Human Cystatin-E Mouse Fetain-A | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction