Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fetuin A/AHSG Antibody (SR2026), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322369100UL
Description
Fetuin A/AHSG Monoclonal antibody specifically detects Fetuin A/AHSG in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Fetuin A/AHSG | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
SR2026 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
A2HS, AHS, alpha-2-HS-glycoprotein, alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, FETUAba-alpha-2-glycoprotein, fetuin-A, HSGA | |
A synthesized peptide derived from human Fetuin A/AHSG (Uniprot #: P02765) | |
100 μg | |
Cancer | |
197 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction