Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fetuin A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA594985
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA594985 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA594985 Supplier Invitrogen™ Supplier No. PA594985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human HCCT tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fetuin A
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene AHSG
Gene Accession No. P02765
Gene Alias 59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; AHSG; alpha 2 HS-glycoprotein alpha 2 (fetuin); alpha 2-HS glycoprotein; alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; asialofetuin; Ba-alpha-2-glycoprotein; BSP; Countertrypin; FETUA; fetuin; fetuin A; FetuinA; fetuin-A; Glycoprotein PP63; HSGA; phosphorylated N-glycoprotein pp63; pp63; PRO2743
Gene Symbols AHSG
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human Fetuin A (46-62aa DYINQNLPWGYKHTLNQ).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.